accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1373920-69-4

IUPAC Name: 3-chloro-5-fluoro-4-methoxy-2-methylbenzaldehyde
Synonyms: 3-chloro-5-fluoro-4-methoxy-2-methylbenzaldehyde
Molecular Weight (g/mol): 202.61
Molecular Formula: C9H8ClFO2
InChi Key: WYIOQVLMXMOGQK-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(C=O)C(C)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results