accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NN

CAS RN 13750-62-4

IUPAC Name: 1-benzyl-2-methyl-1H-imidazole
Synonyms: 1-benzyl-2-methylimidazole
Molecular Weight (g/mol): 172.23
Molecular Formula: C11H12N2
InChi Key: FBHPRUXJQNWTEW-UHFFFAOYSA-N
SMILES: CC1=NC=CN1CC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)