Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Benzyl-2-methylimidazole

Catalog No. 369713500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-971-3500ML 500 mL
36-971-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-971-3500ML Supplier Sigma Aldrich Supplier No. 369713500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-benzyl-2-methylimidazole is a nitrogen heterocycle and forms mixed ligand pt(ii) complexes
  • 1-benzyl-2-methylimidazole on treatement with ammonia and sodium yields 2-methylimidazole
  • Deposition of plasma-polymerized 1-benzyl-2-methylimidazole (ppbmi) thin films onto glass substrates by glow discharge technique have been reported.1-benzyl-2-methylimidazole may be employed as catalyst to investigate the effect of catalysts on the curing reaction of the biphenyl epoxy (4,4α-diglycidyloxy-3,3α,5,5α-tetramethyl biphenyl)dicyclopentadiene type phenolic resin system

Specifications

CAS 13750-62-4
Density 1.105 g/mL (at 25°C (literature))
Boiling Point 125°C to 127°C (3 mmHg)
Molecular Formula C11H12N2
Linear Formula C11H12N2
Refractive Index n20/D 1.5645 (literature)
Quantity 500 mL
MDL Number MFCD00010637
Molecular Weight (g/mol) 172.23
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.