CAS RN 13760-83-3
Erbium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
CAS: 13760-83-3 Molecular Formula: ErF3 Molecular Weight (g/mol): 224.25 MDL Number: MFCD00016074 InChI Key: QGJSAGBHFTXOTM-UHFFFAOYSA-K IUPAC Name: erbium(3+) trifluoride SMILES: [F-].[F-].[F-].[Er+3]
Erbium(III) fluoride, anhydrous, REacton™, 99.99% (REO)
CAS: 13760-83-3 Molecular Formula: ErF3 Molecular Weight (g/mol): 224.25 MDL Number: MFCD00016074 InChI Key: QGJSAGBHFTXOTM-UHFFFAOYSA-K IUPAC Name: erbium(3+) trifluoride SMILES: [F-].[F-].[F-].[Er+3]
MP Biomedicals, Inc Erbium Fluoride, 99.9%, MP Biomedicals
CAS: 13760-83-3 Molecular Formula: ErF3 Molecular Weight (g/mol): 224.25 MDL Number: MFCD00016074 InChI Key: QGJSAGBHFTXOTM-UHFFFAOYSA-K IUPAC Name: erbium(3+) trifluoride SMILES: [F-].[F-].[F-].[Er+3]