Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium Fluoride, 99.9%, MP Biomedicals

Catalog No. ICN203379 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203379 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203379 Supplier MP Biomedicals, Inc Supplier No. 0520337905

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13760-83-3
Molecular Formula ErF3
Molecular Weight (g/mol) 224.25
MDL Number MFCD00016074
InChI Key QGJSAGBHFTXOTM-UHFFFAOYSA-K
IUPAC Name erbium(3+) trifluoride
SMILES [F-].[F-].[F-].[Er+3]

Specifications

Quantity 10 g
Formula Weight 224.28
Chemical Name or Material Erbium Fluoride, 99.9%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.