accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOHHCl

CAS RN 137605-80-2

IUPAC Name: 3-[(morpholin-4-yl)methyl]benzoic acid hydrochloride
Synonyms: 3-(morpholin-4-ylmethyl)benzoic acid hydrochloride
Molecular Weight (g/mol): 257.71
Molecular Formula: C12H16ClNO3
InChi Key: UOMMCPQFAXOMMO-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=CC(CN2CCOCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results