accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CFFF

CAS RN 137644-54-3

IUPAC Name: 4-pentyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)
Synonyms: 4-pentyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 366.51
Molecular Formula: C23H33F3
InChi Key: TXHFUCYJBLLNIG-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1CCC(CC1)C1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results