Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-4'-Pentyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 98.0+%, TCI America™
SDP

Catalog No. P23195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23195G Supplier TCI America Supplier No. P23195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 137644-54-3
Molecular Formula C23H33F3
Molecular Weight (g/mol) 366.512
MDL Number MFCD17019208
InChI Key TXHFUCYJBLLNIG-UHFFFAOYSA-N
Synonym trans,trans-4′C-Amyl-4-(3,4,5-trifluorophenyl)bicyclohexyl
PubChem CID 14984829
IUPAC Name 1,2,3-trifluoro-5-[4-(4-pentylcyclohexyl)cyclohexyl]benzene
SMILES CCCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F

Specifications

Melting Point 87°C
Color White
Quantity 5 g
Formula Weight 366.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4′-Pentyl-4-(3,4,5-trifluorophenyl)bicyclohexyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.