accessibility menu, dialog, popup

UserName

Viewing profile as user:
OClBrFF

CAS RN 1378387-81-5

IUPAC Name: 1-(7-bromo-9,9-difluoro-9H-fluoren-2-yl)-2-chloroethan-1-one
Synonyms: 1-(7-bromo-9,9-difluorofluoren-2-yl)-2-chloroethanone
Molecular Weight (g/mol): 357.58
Molecular Formula: C15H8BrClF2O
InChi Key: KHQZXVZXRCFXSC-UHFFFAOYSA-N
SMILES: FC1(F)C2=C(C=CC(Br)=C2)C2=C1C=C(C=C2)C(=O)CCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results