Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromo-2-(chloroacetyl)-9,9-difluorofluorene 98.0+%, TCI America™
SDP

Catalog No. B51301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B51301G Supplier TCI America Supplier No. B51301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1378387-81-5
Molecular Formula C15H8BrClF2O
Molecular Weight (g/mol) 357.579
MDL Number MFCD28400046
InChI Key KHQZXVZXRCFXSC-UHFFFAOYSA-N
PubChem CID 59611258
IUPAC Name 1-(7-bromo-9,9-difluorofluoren-2-yl)-2-chloroethanone
SMILES C1=CC2=C(C=C1C(=O)CCl)C(C3=C2C=CC(=C3)Br)(F)F

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 1 g
Formula Weight 357.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromo-2-(chloroacetyl)-9,9-difluorofluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.