accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1378825-86-5

IUPAC Name: 1-{2-methyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-2-yl}propan-2-one
Synonyms: 1-{2-methyl-tetrahydrofuro[3,4-d][1,3]dioxol-2-yl}propan-2-one
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: RGYHEPWCPQAJSI-UHFFFAOYNA-N
SMILES: CC(=O)CC1(C)OC2COCC2O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results