accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SiH3CH3CCH3CH3OOOHOH

CAS RN 137915-37-8

IUPAC Name: 2-({[tris(propan-2-yl)silyl]oxy}methyl)-3,4-dihydro-2H-pyran-3,4-diol
Synonyms: 2-{[(triisopropylsilyl)oxy]methyl}-3,4-dihydro-2H-pyran-3,4-diol
Molecular Weight (g/mol): 302.49
Molecular Formula: C15H30O4Si
InChi Key: VYZHAYQIZWKLNP-UHFFFAOYNA-N
SMILES: CC(C)[Si](OCC1OC=CC(O)C1O)(C(C)C)C(C)C
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results