Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-O-(Triisopropylsilyl)-D-glucal 95.0+%, TCI America™
SDP

Catalog No. T1936100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1936100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1936100MG Supplier TCI America Supplier No. T1936100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 137915-37-8
Molecular Formula C15H30O4Si
Molecular Weight (g/mol) 302.486
InChI Key VYZHAYQIZWKLNP-KFWWJZLASA-N
Synonym 6-o-triisopropylsilyl-d-glucal, 2r,3s,4r-2-triisopropylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diol, 1,5-anhydro-2-deoxy-6-o-triisopropylsilyl-d-arabino-hex-1-enitol, 6-o-triisopropylsilyl-1,2-didehydro-1,2-dideoxy-d-glucopyranose
PubChem CID 10614345
IUPAC Name (2R,3S,4R)-2-[tri(propan-2-yl)silyloxymethyl]-3,4-dihydro-2H-pyran-3,4-diol
SMILES CC(C)[Si](C(C)C)(C(C)C)OCC1C(C(C=CO1)O)O

Specifications

Quantity 100 mg
Formula Weight 302.49
Percent Purity ≥95.0% (GC)
Chemical Name or Material 6-O-(Triisopropylsilyl)-D-glucal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.