accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CH3CCH3SiCH3H3CCH3CH3

CAS RN 1379822-08-8

IUPAC Name: (2-{2,4-dimethyl-5-[2-(trimethylsilyl)ethynyl]phenyl}ethynyl)trimethylsilane
Synonyms: (2-{2,4-dimethyl-5-[2-(trimethylsilyl)ethynyl]phenyl}ethynyl)trimethylsilane
Molecular Weight (g/mol): 298.58
Molecular Formula: C18H26Si2
InChi Key: GTCZWWHSXWEQGR-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=C1C#C[Si](C)(C)C)C#C[Si](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results