Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dimethyl-2,4-bis[2-(trimethylsilyl)ethynyl]benzene 97.0+%, TCI America™
SDP

Catalog No. D50295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50295G 5 g
D50291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50295G Supplier TCI America Supplier No. D50295G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1379822-08-8
Molecular Formula C18H26Si2
Molecular Weight (g/mol) 298.576
InChI Key GTCZWWHSXWEQGR-UHFFFAOYSA-N
PubChem CID 131637290
IUPAC Name 2-[2,4-dimethyl-5-(2-trimethylsilylethynyl)phenyl]ethynyl-trimethylsilane
SMILES CC1=CC(=C(C=C1C#C[Si](C)(C)C)C#C[Si](C)(C)C)C

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 298.58
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dimethyl-2,4-bis[2-(trimethylsilyl)ethynyl]benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.