accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138006-41-4

IUPAC Name: 3-bromo-2,5-dichloropyridine
Synonyms: 3-bromo-2,5-dichloropyridine
Molecular Weight (g/mol): 226.88
Molecular Formula: C5H2BrCl2N
InChi Key: CDBHWRZEXFQNBI-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results