accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHNONH3C

CAS RN 138046-43-2

IUPAC Name: 1-cycloheptyl-1-[(9H-fluoren-2-yl)methyl]-3-(2,4,6-trimethylphenyl)urea
Synonyms: 1-cycloheptyl-1-(9H-fluoren-2-ylmethyl)-3-(2,4,6-trimethylphenyl)urea
Molecular Weight (g/mol): 452.64
Molecular Formula: C31H36N2O
InChi Key: FMLJREWZCZHGGW-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(NC(=O)N(CC2=CC3=C(C=C2)C2=CC=CC=C2C3)C2CCCCCC2)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results