accessibility menu, dialog, popup

UserName

Viewing profile as user:
CFFZnBr

CAS RN 1380918-98-8

IUPAC Name: 2,6-difluorobenzen-1-ide; bromozincylium
Synonyms: 2,6-difluorobenzen-1-ide; bromozincylium
Molecular Weight (g/mol): 258.37
Molecular Formula: C6H3BrF2Zn
InChi Key: PYSMUDJGPPCOFC-UHFFFAOYSA-M
SMILES: [Zn+]Br.FC1=CC=CC(F)=[C-]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results