Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58845AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58845AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58845AD Supplier Thermo Scientific Chemicals Supplier No. H58845AD
Add to Cart

CAS: 1380918-98-8 | C6H3BrF2Zn | 258.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1380918-98-8
Molecular Formula C6H3BrF2Zn
Molecular Weight (g/mol) 258.37
MDL Number MFCD09801448
InChI Key PYSMUDJGPPCOFC-UHFFFAOYSA-M
Synonym 2,6-difluorophenylzinc bromide, bromo 2,6-difluorophenyl zinc, 2,6-difluorophenylzinc bromide solution, 2,6-difluorophenylzinc bromide 0.5m in tetrahydrofuran, 2,6-difluorophenylzinc bromide solution, 0.5 m in thf, 2,6-difluorophenylzinc bromide, 0.5m in tetrahydrofuran
PubChem CID 24722677
IUPAC Name 2,6-difluorobenzen-1-ide; bromozincylium
SMILES [Zn+]Br.FC1=CC=CC(F)=[C-]1

Specifications

Density 1.006
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Formula Weight 258.37
Chemical Name or Material 2,6-Difluorophenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.