accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13813-22-4

IUPAC Name: lanthanum(3+) triiodide
Synonyms: lanthanum(3+) triiodide
Molecular Weight (g/mol): 519.62
Molecular Formula: I3La
InChi Key: KYKBXWMMXCGRBA-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[La+3]
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (4)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (4)
  • (2)