CAS RN 13813-43-9
IUPAC Name:
thulium(3+) triiodide
Synonyms:
thulium(3+) triiodide
Molecular Weight (g/mol):
549.65
Molecular Formula:
I3Tm
InChi Key:
LZOMHYVAEHYDST-UHFFFAOYSA-K
SMILES:
[I-].[I-].[I-].[Tm+3]
1
–
2
of
2
results
1
Thulium(III) iodide, 99.9% (REO), Thermo Scientific Chemicals
CAS: 13813-43-9 MDL Number: MFCD00050141 Synonym: thulium iii iodide,acmc-1boxt
2
Thulium(III) iodide, ultra dry, 99.99% (REO)
CAS: 13813-43-9 MDL Number: MFCD00050141 Synonym: thulium iii iodide,acmc-1boxt