accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13816-04-1

H3CCH2NS

CAS RN 13816-04-1

IUPAC Name: 2-(prop-1-en-2-yl)-1,3-thiazole
Synonyms: 2-(prop-1-en-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 125.19
Molecular Formula: C6H7NS
InChi Key: UDPXHYZDXKTGLD-UHFFFAOYSA-N
SMILES: CC(=C)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results