Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropenylthiazole (stabilized with HQ) 98.0+%, TCI America™
SDP

Catalog No. I047525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I047525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I047525G Supplier TCI America Supplier No. I047525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13816-04-1
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.189
MDL Number MFCD00671547
InChI Key UDPXHYZDXKTGLD-UHFFFAOYSA-N
PubChem CID 580455
IUPAC Name 2-prop-1-en-2-yl-1,3-thiazole
SMILES CC(=C)C1=NC=CS1

Specifications

Boiling Point 63°C
Quantity 25 g
Formula Weight 125.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropenylthiazole (stabilized with HQ)
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.