accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13820-09-2

IUPAC Name: 1,1,1-trimethoxypentane
Synonyms: 1,1,1-trimethoxypentane
Molecular Weight (g/mol): 162.23
Molecular Formula: C8H18O3
InChi Key: XUXVVQKJULMMKX-UHFFFAOYSA-N
SMILES: CCCCC(OC)(OC)OC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)