accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13823-29-5

IUPAC Name: thorium(4+) tetranitrate
Synonyms: thorium(4+) tetranitrate
Molecular Weight (g/mol): 480.05
Molecular Formula: N4O12Th
InChi Key: VGBPIHVLVSGJGR-UHFFFAOYSA-N
SMILES: [Th+4].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results