Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thorium AA Standard, 1000 ppm Th in 3% HNO3, Ricca Chemical
SDP

Catalog No. ATH1KN500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ATH1KN500 500 mL Natural Poly Bottle
ATH1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ATH1KN500 Supplier Ricca Chemical Company Supplier No. ATH1KN500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13823-29-5 , 7697-37-2 , 7732-18-5

Specifications

Packaging Natural Poly Bottle
CAS Min % 94.0, 4.22, 0.23
CAS Max % 97.0, 4.39, 0.23
Odor Odorless
Color Colorless
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg Th (1, 000ppm Th)Th(NO3)4 in 3% HNO3
Chemical Name or Material Thorium AA Standard
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.