CAS RN 13825-76-8
CAS RN 13825-76-8
IUPAC Name:
tribromoholmium
Synonyms:
tribromoholmium
Molecular Weight (g/mol):
404.64
Molecular Formula:
Br3Ho
InChi Key:
MZNSYJWLQLXLHE-UHFFFAOYSA-K
SMILES:
Br[Ho](Br)Br
1
–
1
of
1
results
1
Thermo Scientific Chemicals Holmium(III) bromide, ultra dry, 99.98% (REO), Thermo Scientific™
CAS: 13825-76-8 MDL Number: MFCD00049744