accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiLiNiBrBrBrBr

CAS RN 13826-95-4

IUPAC Name: dilithium(1+) tetrabromonickeldiuide
Synonyms: dilithium(1+) tetrabromonickelate(2-)
Molecular Weight (g/mol): 392.19
Molecular Formula: Br4Li2Ni
InChi Key: AVJDNPHXGKKFIA-UHFFFAOYSA-J
SMILES: [Li+].[Li+].Br[Ni--](Br)(Br)Br
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results