Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™

Catalog No. AC434548000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434548000 800 mL AcroSeal™ Glass Bottle
AC434541000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434548000 Supplier Thermo Scientific Chemicals Supplier No. 434548000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 13826-95-4 | Br4Li2Ni | 392.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-95-4 , 109-99-9
Molecular Formula Br4Li2Ni
Molecular Weight (g/mol) 392.19
InChI Key AVJDNPHXGKKFIA-UHFFFAOYSA-J
Synonym di-lithium tetrabromonickelate ii, agn-pc-0nf0dx, lithium tetrabromonickelate ii solution, dilithium tetrabromonickelate ii solution
PubChem CID 11047451
IUPAC Name dilithium;tetrabromonickel(2-)
SMILES [Li+].[Li+].[Ni-2](Br)(Br)(Br)Br

Specifications

Color Blue-Green to Brown
Physical Form Solution
Quantity 800 mL
Linear Formula Li2NiBr4
Flash Point −18°C
Formula Weight 392.19
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.065
Density 1.0650g/mL
Chemical Name or Material Dilithium tetrabromonickelate(II)
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
May cause cancer.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.