accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138261-41-3

IUPAC Name: 2-chloro-5-{[2-(nitroamino)-4,5-dihydro-1H-imidazol-1-yl]methyl}pyridine
Synonyms: imidacloprid
Molecular Weight (g/mol): 255.66
Molecular Formula: C9H10ClN5O2
InChi Key: YWTYJOPNNQFBPC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)NC1=NCCN1CC1=CC=C(Cl)N=C1
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)