accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1383806-53-8

IUPAC Name: 2-(5-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(5-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 252.09
Molecular Formula: C13H18BFO3
InChi Key: HFSNSSDHOJSZKI-UHFFFAOYSA-N
SMILES: COC1=C(C=C(F)C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results