accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONNNHNN

CAS RN 138402-11-6

IUPAC Name: 2-butyl-3-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1,3-diazaspiro[4.4]non-1-en-4-one
Synonyms: irbesartan bristol myers brand of irbesartan sanofi winthrop brand of irbesartan bms brand of irbesartan irbesartan (substance) avalide aprovel irbesartan preparation irbesartan (product) 厄贝沙坦
Molecular Weight (g/mol): 428.54
Molecular Formula: C25H28N6O
InChi Key: YOSHYTLCDANDAN-UHFFFAOYSA-N
SMILES: CCCCC1=NC2(CCCC2)C(=O)N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)