accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138449-07-7

IUPAC Name: (2S)-N-benzyl-2-{[(2S,4R)-4-hydroxy-1-(1-methyl-1H-indole-3-carbonyl)pyrrolidin-2-yl]formamido}-N-methyl-3-(naphthalen-2-yl)propanamide
Synonyms: (2S)-N-benzyl-2-{[(2S,4R)-4-hydroxy-1-(1-methylindole-3-carbonyl)pyrrolidin-2-yl]formamido}-N-methyl-3-(naphthalen-2-yl)propanamide
Molecular Weight (g/mol): 588.71
Molecular Formula: C36H36N4O4
InChi Key: BFNKQTIJVFGCKQ-PDJGWCFMSA-N
SMILES: CN(CC1=CC=CC=C1)C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)[C@@H]1C[C@@H](O)CN1C(=O)C1=CN(C)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results