accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13845-36-8

IUPAC Name: pentapotassium bis(phosphonatooxy)phosphinate
Synonyms: pentapotassium triphosphate
Molecular Weight (g/mol): 448.4
Molecular Formula: K5O10P3
InChi Key: ATGAWOHQWWULNK-UHFFFAOYSA-I
SMILES: [K+].[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)