Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Tripolyphosphate, Spectrum™ Chemical
SDP

Catalog No. 18605744 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-744 45 kg Poly Bottle
18-605-741 500 g Poly Bottle
18-605-742 2.5 kg Poly Bottle
18-605-743 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-744 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P166945KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1669, 13845-36-8, K5P3O10

Spectrum™ Chemical Potassium Tripolyphosphate

Chemical Identifiers

CAS 13845-36-8
Molecular Formula K5O10P3
Molecular Weight (g/mol) 448.40
InChI Key ATGAWOHQWWULNK-UHFFFAOYSA-I
IUPAC Name pentapotassium bis(phosphonatooxy)phosphinate
SMILES [K+].[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 45 kg
Grade Ungraded
Packaging Poly Bottle
Chemical Name or Material Potassium Tripolyphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.