accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOHO

CAS RN 138485-81-1

IUPAC Name: 3-[4-(methylsulfanyl)phenyl]propanoic acid
Synonyms: 3-[4-(methylsulfanyl)phenyl]propanoic acid
Molecular Weight (g/mol): 196.26
Molecular Formula: C10H12O2S
InChi Key: ASTVFRXZLBGVRL-UHFFFAOYSA-N
SMILES: CSC1=CC=C(CCC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results