accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOCH2

CAS RN 138551-10-7

IUPAC Name: 1,4-bis(undec-10-en-1-yloxy)benzene
Synonyms: 1,4-bis(undec-10-en-1-yloxy)benzene
Molecular Weight (g/mol): 414.67
Molecular Formula: C28H46O2
InChi Key: WCFSIYFISCSTNX-UHFFFAOYSA-N
SMILES: C=CCCCCCCCCCOC1=CC=C(OCCCCCCCCCC=C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results