Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(10-undecenyloxy)benzene 97.0+%, TCI America™
SDP

Catalog No. B35751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35751G Supplier TCI America Supplier No. B35751G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138551-10-7
Molecular Formula C28H46O2
Molecular Weight (g/mol) 414.674
InChI Key WCFSIYFISCSTNX-UHFFFAOYSA-N
PubChem CID 22979406
IUPAC Name 1,4-bis(undec-10-enoxy)benzene
SMILES C=CCCCCCCCCCOC1=CC=C(C=C1)OCCCCCCCCCC=C

Specifications

Melting Point 61°C
Color White
Boiling Point 160°C
Quantity 1 g
Formula Weight 414.67
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(10-undecenyloxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.