CAS RN 13859-68-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hexaamminenickel(II) iodide, 99.999% (metals basis), Thermo Scientific™
CAS: 13859-68-2 Molecular Formula: Ni(NH3)6I2 MDL Number: MFCD00058808 Synonym: diiodonickel-ammonia 1/6