accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONSCH3

CAS RN 1386647-53-5

IUPAC Name: ethyl 2-cyclopropyl-4-methyl-1,3-thiazole-5-carboxylate
Synonyms: ethyl 2-cyclopropyl-4-methyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 211.28
Molecular Formula: C10H13NO2S
InChi Key: BMOWAHYIBJQJIU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(S1)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results