accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3NOOH3CH3CCH3

CAS RN 1387445-50-2

IUPAC Name: tert-butyl 5-methoxy-7-methyl-1H-indole-1-carboxylate
Synonyms: tert-butyl 5-methoxy-7-methylindole-1-carboxylate
Molecular Weight (g/mol): 261.32
Molecular Formula: C15H19NO3
InChi Key: OMZYGXJHKONVSJ-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C2N(C=CC2=C1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results