Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-5-methoxy-7-methylindole, 97%, Thermo Scientific™

Catalog No. AAH63905MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63905MD 250 mg
AAH6390503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63905MD Supplier Thermo Scientific Chemicals Supplier No. H63905MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1387445-50-2
Molecular Formula C15H19NO3
Molecular Weight (g/mol) 261.321
InChI Key OMZYGXJHKONVSJ-UHFFFAOYSA-N
Synonym tert-butyl 5-methoxy-7-methyl-1h-indole-1-carboxylate, n-boc-5-methoxy-7-methylindole
PubChem CID 100913508
IUPAC Name tert-butyl 5-methoxy-7-methylindole-1-carboxylate
SMILES CC1=C2C(=CC(=C1)OC)C=CN2C(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 261.32
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Boc-5-methoxy-7-methylindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.