CAS RN 13877-91-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
Ricca Chemical Company Ocimene, Natural Isolate, Ricca Chemical
Ocimene, Natural Isolate is a natural isolate food additive.