accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138775-48-1

IUPAC Name: (2R)-3-[4-(benzyloxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2R)-3-[4-(benzyloxy)phenyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 493.56
Molecular Formula: C31H27NO5
InChi Key: REHSJSKPWIOKIJ-KSTXGFNDNA-N
SMILES: OC(=O)[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)