Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-O-benzyl-D-tyrosine, 95%, Thermo Scientific™

Catalog No. AAH6659903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6659903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6659903 Supplier Thermo Scientific Chemicals Supplier No. H6659903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138775-48-1
Molecular Formula C31H27NO5
Molecular Weight (g/mol) 493.56
MDL Number MFCD00235837
InChI Key REHSJSKPWIOKIJ-KSTXGFNDNA-N
Synonym fmoc-d-tyr bzl-oh, fmoc-o-benzyl-d-tyr, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, 2r-3-4-benzyloxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-o-benzyl-l-tyr, n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-d-tyrosine, o-benzyl-n-9h-fluoren-9-yl methoxy carbonyl-d-tyrosine, d-tyrosine,n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl
PubChem CID 40428316
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid
SMILES OC(=O)[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White
Quantity 1 g
Formula Weight 439.56
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-O-benzyl-D-tyrosine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.