accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONNFNH2

CAS RN 138907-68-3

IUPAC Name: ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Synonyms: ethyl 5-amino-1-(4-fluorophenyl)pyrazole-4-carboxylate
Molecular Weight (g/mol): 249.25
Molecular Formula: C12H12FN3O2
InChi Key: RPPPCKSHIYWAPW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)N(N=C1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results