Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate, 97%, Thermo Scientific™

Catalog No. RH00119EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RH00119EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RH00119EA Supplier Thermo Scientific Chemicals Supplier No. RH00119EA
Add to Cart

Chemical Identifiers

CAS 138907-68-3
Molecular Formula C12H12FN3O2
Molecular Weight (g/mol) 249.25
MDL Number MFCD00173917
InChI Key RPPPCKSHIYWAPW-UHFFFAOYSA-N
Synonym ethyl 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluorophenyl pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluoro-phenyl-1h-pyrazole-4-carboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylic acid ethyl ester, 5-amino-4-ethoxycarbonyl-1-4-fluorophenyl-1h-pyrazole, 1-5-amino-4-ethoxycarbonyl-1h-pyrazol-1-yl-4-fluorobenzene, maybridge1_006170, ethyl5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethylaminofluorophenylpyrazolecarboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxyli
PubChem CID 688689
IUPAC Name ethyl 5-amino-1-(4-fluorophenyl)pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(N)N(N=C1)C1=CC=C(F)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 249.24
Percent Purity 97%
Chemical Name or Material ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.