accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138926-19-9

IUPAC Name: sodium hydrate hydrogen {1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropyl}phosphonate
Synonyms: sodium hydrate hydrogen 1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropylphosphonate
Molecular Weight (g/mol): 359.23
Molecular Formula: C9H24NNaO8P2
InChi Key: VBDRTGFACFYFCT-UHFFFAOYNA-M
SMILES: O.[Na+].CCCCCN(C)CCC(O)(P(O)(O)=O)P(O)([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results