Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ibandronate sodium salt monohydrate

Catalog No. AC463150050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463150050 5 g
AC463150010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463150050 Supplier Thermo Scientific Chemicals Supplier No. 463150050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 138926-19-9 | C9H24NNaO8P2 | 359.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138926-19-9
Molecular Formula C9H24NNaO8P2
Molecular Weight (g/mol) 359.23
InChI Key VBDRTGFACFYFCT-UHFFFAOYNA-M
IUPAC Name sodium hydrate hydrogen {1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropyl}phosphonate
SMILES O.[Na+].CCCCCN(C)CCC(O)(P(O)(O)=O)P(O)([O-])=O

Specifications

Melting Point 175.0°C to 177.0°C
Quantity 5 g
Formula Weight 359.23
Chemical Name or Material Ibandronate sodium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.