accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CSOHOO

CAS RN 13893-34-0

IUPAC Name: octadecane-1-sulfonic acid sodium
Synonyms: octadecane-1-sulfonic acid sodium
Molecular Weight (g/mol): 357.55
Molecular Formula: C18H38NaO3S
InChi Key: NIMUYFTYMXTPAV-UHFFFAOYSA-N
SMILES: [Na].CCCCCCCCCCCCCCCCCCS(O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)